3M™ Powerflow™ Face-Mounted Powered Air Purifying Respirators

Ideal for lead, asbestos and other hazardous particulates

Manufacturer: 3M™ 6800PF

 View more versions of this product

Catalog No. 19-096-897

  Check Availability
Add to Cart

    Specifications

    Certifications ANSI Z87.1-2003
    Filter Type HE Filter
    For Use With (Application) Lead, asbestos and hazardous particulate
    Includes Medium full facepiece, motor blower unit, battery pack, flow meter
    Mounting Type Face
    Size Medium
    Weight (English) 6 lb.
    Weight (Metric) 2.72kg
    Airflow Rate 4cfm
    Product Type Powered Air Purifying Respirator

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit