Learn More
3M™ H-700 Series Cap Style Hard Hats with 4-Point Ratchet Suspension
Protect top of the head due to potential hazard of small falling objects or exposed fixed objects
Supplier: 3M™ 7000030066
Description
- Suspension adjusts easily to allow for customized height and fit
- Replaceable brow pad for more comfort and extended shelf life
- Short brim for upward visibility
- Meets requirements of ANSI/ISEA Z89.1-2009 Type 1, Class C
- ANSI/ISEA compliant when worn in the forward and reverse donning positions
- Available in various colors
- 3M Catalog# H-701R, H-702R, H-703R, H-704R, H-705R, H-706R, H-707R, H-708R, H-709R, H-710R, H-711R, H-712R, H-713R
Specifications
Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E | |
Standard brow pad | |
Cap Style Hard Hat | |
4-Point Ratchet |
Bright Yellow | |
H-709R | |
20/Cs. |