3M™ H-700 Series Cap Style Hard Hats with 4-Point Ratchet Suspension
Protect top of the head due to potential hazard of small falling objects or exposed fixed objects
Manufacturer: 3M™ H703R
 View more versions of this product
Catalog No. 17-623-313
  Check Availability
Add to Cart
    Specifications
    Product Type Cap Style Hard Hat
    Includes Standard brow pad
    Certifications/Compliance Meets ANSI/ISEA Z89.1-2009 Type 1, Class C, G and E
    Color Blue
    Quantity 20/Cs.
    Suspension Type 4-Point Ratchet
  • Description & Specifications
Related Products
Product Content Feedback