Learn More
3M™ QuickLatch™ Respirators Cartridges and Filters
Manufacturer:  3M™ 5052700000
 View more versions of this product
Catalog No. 19-063-506
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Low profile filters, cartridges, and cartridge/filter combinations have off-center adapters for maximum peripheral vision. Conform™ flexible bayonet-style filter is extremely lightweight. Plastic cartridges are lightweight and offer varied levels of protection. Labels are affixed to the side of the cartridge and are easy to read so that the proper cartridge is used for the contaiminant. For use with all AOSafety 8000 Series respirators.
Order Info
10 per pack.
Compliance
NIOSH approved.
Specifications
Specifications
Safety
50527-00000