Learn More
3M™ QuickLatch™ Respirators Cartridges and Filters
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-063-506
View Documents
3M™
5052700000
N/A Organic Vapor/Acid Gas Cartridge This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Low profile filters, cartridges, and cartridge/filter combinations have off-center adapters for maximum peripheral vision. Conform™ flexible bayonet-style filter is extremely lightweight. Plastic cartridges are lightweight and offer varied levels of protection. Labels are affixed to the side of the cartridge and are easy to read so that the proper cartridge is used for the contaiminant. For use with all AOSafety 8000 Series respirators.
Order Info
10 per pack.
Compliance
NIOSH approved.