3M™ QuickLatch™ Respirators Cartridges and Filters
Manufacturer: 3M™ 50537-00000
 View all options for this product
Catalog No. 19-063-526
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Low profile filters, cartridges, and cartridge/filter combinations have off-center adapters for maximum peripheral vision. Conform™ flexible bayonet-style filter is extremely lightweight. Plastic cartridges are lightweight and offer varied levels of protection. Labels are affixed to the side of the cartridge and are easy to read so that the proper cartridge is used for the contaiminant. For use with all AOSafety 8000 Series respirators.
  • Description & Specifications
Related Products
Product Content Feedback