Learn More
3M™ Refine™ Eyewear for Women
Specially sized for the female facial profile
Manufacturer:  3M™ 117390000020
 View more versions of this product
Catalog No. 19-065-414
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • Fits closer and more proportionally to the eyes, cheeks and brow
  • Small, lightweight frame for full-shift comfort
  • Wraparound lens for optimum orbital protection, slim frame design
  • Padded, adjustable nose bridge available
  • Teal frame
  • 3M Catalog# 11735-00000-20
Compliance
Meets ANSI Z87.1-2003 standards.
Specifications
Specifications
Meets ANSI Z87.1- 2010 standards.
Mocha
Anti-fog
20/cs.
Safety
Gray
Refine 102