3M™ Replacement Parts for Air-Mate™ Belt Mounted High Efficiency PAPR

Replacement parts for Air-Mate high efficiency powered air purifying respirator systems

Manufacturer: 3M™ 5200363R01

 View more versions of this product

Catalog No. 18-033

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) 3M™ air-mate™ high efficiency powered air purifying respirator system
    Product Type Air Purifying Respirator Unit
    For Use With (Application) Respiratory protection against certain airborne particulate contaminants including dusts, fumes, mists, radionuclides and asbestos
    Includes Built-in fuse and an on/off switch
    Mounting Type Belt
    Weight (Metric) 0.5kg
    Weight (English) 1.1 lb.
    Certifications NIOSH
    Battery Type NiCd (rechargeable)
    Battery Life 8 hr.
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit