Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Repositionable 75 Spray Adhesive

Adheres in seconds, but stays tacky for repositioning

Supplier:  3M™ 30067

Catalog No. 19-047-240

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Provides tape-like bonds on many lightweight materials, adhering in seconds yet preserving an extra-long tack range that allows lifting and repositioning of materials.

  • Excellent for production line work where parts are held in place temporarily prior to final assembly
  • Bonds paper, cardboard, acetate, foil, fabrics, sheeting, and similar lightweight materials
  • Will not bleed through, stain, or wrinkle most materials
  • Clear in color.

Order Info

16 fl. oz. (473mL) can.

Specifications

Specifications

Spray Adhesive
Clear
12 Pk.
76.66°C
473.17 mL
Bonds paper, cardboard, acetate, foil, fabrics, sheeting, and similar lightweight materials
Aerosol Can
170°F
16 oz.
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

ShelfLife : 15 Months

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit