Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Versaflo™ Breathing Tubes for Headtops and Air Delivery Units
Lightweight BT-series breathing tubes are compatible with all 3M™ Versaflo™ headtops and air delivery units.
Supplier: 3M™ 7100134625
Description
- Long, lightweight breathing tube that is easy to wipe down
- Quick release swivel enables one-handed connection and release
- Disposable breathing tube cover is available to help cleaning and decontamination operations
- 3M Catalog# BT-20S and BT-20L
Specifications
Breathing Tube | |
Assembly, facility maintenance, grinding, machine operation, painting, sanding | |
BT-20S | |
71.1 cm |
Small/Medium | |
Versaflo™ PAPR Systems | |
28 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction