3M™ S-Series Integrated Suspension Hoods and Head Covers

Lightweight, loose-fitting hoods and head Covers offer both comfort and protection

Manufacturer: 3M™ S8075

 View more versions of this product

Catalog No. 19-805-542

  Check Availability
Add to Cart

    Specifications

    Quantity 5 Case
    For Use With (Equipment) 3M™ premium head suspension of 3M™ powered air purifying and supplied air respirator system,3M™ premium head suspension of 3M™ powered air purifying and supplied air respirator system
    Product Type Replacement hood with sealed seams and inner shroud
    For Use With (Application) Assembly, facility maintenance, machine operation, painting, sanding
    Includes Sealed Seams and inner shroud
    Material Kappler™ Zytron™ 200 polypropylene fabric
    Size Standard
    Certifications NIOSH
    Color Gray
    Seam Type Sealed

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit