Learn More
3M™ S-Series Premium Suspension Hoods and Replacements
Lightweight, loose-fitting hoods and head Covers offer both comfort and protection
Manufacturer:  3M™ 7000127432
 View more versions of this product
Catalog No. 19-805-600
Add to cart
Description
Description
Integrated Suspension Hoods and Head Covers
  • Head covers provide head and face coverage; hoods cover head, face, and neck
  • Fully assembled and ready to use
  • Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or situtations where there is less soiling of the outer fabric
  • Integrated models feature wide faceseal with soft elastic for improved comfort and reduced noise
  • Premium Reusable Suspensions with Hoods
    • Ideal for high disposal rate environments
    • Soiled hood is replaced; suspension and air ducting are reused
    • Rigid ducting for consistent air path
    • Externally adjustable airflow control allows wearer to modify airflow distribution
    • Suspension features independent adjustments for the crown and head circumference; allows custom fit for wide range of head sizes
    • 3M Catalog# S-133L-5, S-133S-5, S-433S-5, S-533L, S-533S, S-707-10, S-757, S-805-5, S-807-5, S-857
Specifications
Specifications
White
Assembly, facility maintenance, machine operation, painting, sanding, pharmaceutical manufacturing, liquid handling, battery manufacturing and recycling, petrochemical refining and extraction
8.19 in.
Head and face coverage. Integrated Comfort suspension; General purpose, cost-effective low-linting fabric
25.4cm
1 Case
9.63 in.
Hood
10 x 9.63 x 8.19 in. (25.4 x 24.46 x 20.8cm)
3M™ powered air purifying respirator and supplied air respirator valves
20.8cm
10 in.
Polyurethane Coated Knitted Polyamide
Small, Medium
24.46cm
Safety