Learn More
3M™ Scotch™ Vinyl Electrical Color-Coding Tape 35
Tough, premium-grade, 7mil vinyl
Manufacturer:  3M™ 7000058438
 View more versions of this product
Catalog No. 19-047-285
Add to cart
Description
Description
  • Stretches to conform to virtually any shape that requires color-coded insulation
  • Provides joint, termination, and connector insulation
  • Stands up to harsh weather conditions and broad temperature ranges
  • 3M Catalog# 35-1/2X20FT-BR , 3M Catalog# 35-1/2X20FT-OR , 3M Catalog# 35-1/2X20FT-RD
Order Info
Rolls measure 1/2 in. x 20 ft.. Made in USA.
Compliance
UL Listed.
Specifications
Specifications
Safety
Orange
Premium
20 ft.
5 Year(s)
0°C to 105°C
7 mil
Tape
Electrical Sealing Tape
Resin, Rubber
CSA, UL, VDE 0340-TEIL1, RoHS
Phase identification and color coding of motor leads and piping system
609.6 cm
32 to 220°F
17 lb./in.
0.5 in.
PVC
10 Roll(s)