Learn More
3M™ Sealed Seam Hood Assemblies
Used on 3M™ Belt-Mounted Powered Air Purifying Respirator (PAPR)s and Supplied Air Respirator Systems
Manufacturer:  3M™ H611
 View more versions of this product
Catalog No. 19-038-907
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
White
Sealed Seam Hood Assembly
w/Collar and cap suspension
Standard
Pinlock
Belt mounted powered air purifying respirator and supplied air respirator system
Pharmaceutical, machine operation
Tychem™ SL
NIOSH
Accessories