3M™ Speedglas™ FlexView Helmet with SideWindows and ProTop System
Increase welding performance with improved visibility, safety and comfort
Manufacturer: 3M™ 04-1516-21SW
 View all options for this product
Catalog No. 19-150-0534
  Check Availability
Add to Cart
    Product Type Welding Helmet
    For Use With (Application) Ideal to increase welding performance with improved visibility, safety and comfort
    Includes Variable filter 9-13; With SideWindows
    Length (English) Viewing Area 2.13 in.
    Length (Metric) Viewing Area 5.4cm
    • Clear polycarbonate faceshield for grinding
    • ProTop hard hat with special pivot that allows the shield to nest down on hard hat for better balance
    • Constant UV/IR protection
    • Helmet option with side windows to increase peripheral viewing area
    • Features exhaust vents to help reduce stuffiness
  • Description & Specifications
Related Products
Product Content Feedback