3M™ Speedglas™ FlexView Helmet with SideWindows and ProTop System
Increase welding performance with improved visibility, safety and comfort
Manufacturer: 3M™ 04-1516-00
 View all options for this product
Catalog No. 19-150-0519
  Check Availability
Add to Cart
    Product Type Welding Helmet
    For Use With (Application) Ideal to increase welding performance with improved visibility, safety and comfort
    Includes No ADF Filter
    Weight (Metric) 1.7kg
    Weight (English) 3.73 lb.
    Length (English) Viewing Area N/A
    Length (Metric) Viewing Area N/A
    • Clear polycarbonate faceshield for grinding
    • ProTop hard hat with special pivot that allows the shield to nest down on hard hat for better balance
    • Constant UV/IR protection
    • Helmet option with side windows to increase peripheral viewing area
    • Features exhaust vents to help reduce stuffiness
  • Description & Specifications
Related Products
Product Content Feedback