Learn More
3M™ Speedglas™ Fresh-Air III Supplied Air System
Supplier: 3M™ 27570220SW
Description
Complete respiratory system and helmet included. Speedglas™ Fresh-Air III Supplied Air Systems can provide overhead protection, increased peripheral vision, excellent optical quality and function. Storage bag, welding cap and starter kit included.Supplied air respiratory systems:V-100 Vortex air-cooling valve, belt, breathing tube, complete helmet assembly with ADFV-300 air-regulating valve, belt, breathing tube, complete helmet assembly with ADFV-100SR cool-air valve with Schrader fitting. Auto-Darkening Filter not includedLightweight belt-mounted valve permits welders to lower temperature of air entering headpiece by up to 50°F (28°C), ideal for hot, strenuous jobsThe 3M™ Speedglas™ Welding Helmet 9100 MP features:- Flip-up assembly for grinding and prep work
- SideWindows - Shade 5 side filters that increase peripheral vision
- Ergonomic, adjustable head suspension for increased comfort, helmet stability and weight distribution
- 3M™ Speedglas™ Auto-Darkening Filter 9100X with 2.1 x 4.2 in. viewing area
- Selectable Shades 5, 8-13 and Shade 3 light state; Grinding and Tack modes
- For use with all Stick, MIG, and TIG welding processes
- Five-position sensitivity control
- Adjustable recovery delay
- Offers Assigned Protection Factor (APF) of 25 when used as a NIOSH-approved system
- Meets ANSI Z89.1 - 2009 requirements for Type 1 Class G hardhats
- Meets ANSI Z87.1-2010
- Earmuff protection available as optional accessory
Specifications
4.2 in. | |
For weld preparations by providing five levels of welder protection such as head, eye, face, respiratory and optional hearing combined into one system | |
27-5702-20SW |
NIOSH, OSHA, CSA Z94.3 for CSA, ANSI Z87.1 - 2010 | |
V-100 cooling valve; With 3M Speedglas welding filter 9100X |