3M™ Speedglas™ Fresh-Air III Supplied Air System
Manufacturer: 3M™ 27590220SW
 View more versions of this product
Catalog No. 19-164-117
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For weld preparations by providing five levels of welder protection such as head, eye, face, respiratory and optional hearing combined into one system
    Includes V-300 cooling valve; With 3M Speedglas welding filter 9100X
    Width (English) 3.7 in.
    Certifications NIOSH, OSHA, CSA Z94.3 for CSA, ANSI Z87.1 - 2010,NIOSH, OSHA, CSA Z94.3 for CSA, ANSI Z87.1 - 2010
  • Description & Specifications
Related Products
Product Content Feedback