Learn More
3M™ Speedglas™ Fresh-Air III Supplied Air System
Manufacturer:  3M™ 27570210SW
 View more versions of this product
Catalog No. 19-164-113
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
NIOSH, OSHA, CSA Z94.3 for CSA, ANSI Z87.1 - 2010
For weld preparations by providing five levels of welder protection such as head, eye, face, respiratory and optional hearing combined into one system
V-100 cooling valve; With 3M Speedglas welding filter 9100V
3.7 in.