Learn More
3M™ Speedglas Helmet: Protection Plates
Designed for use with Speedglas Helmets
Supplier: 3M™ 02069000
Description
3M offers a variety of replacement protection plates and accessories designed to protect the filter from spark and spatter.- Replacement inside protection plates for 3M™ Speedglas™ Filters 9002X and 9000X/XF/Xi
- Speedglas 9000 standard outside protection plate
- Replacement silver front panel (only) for 3M™ Speedglas™ FlexView filter assembly
- Replacement face seal for 3M™ Speedglas™ Helmet 9000 HWR
- Throat protector with Velcro Fasteners attaches to 3M™ Speedglas™ Helmets 9000 Series
- Leather throat and side protectors for 3M™ Speedglas™ Fresh-air (FA) PA and SA (APC style helmet)
- 3M Catalog# 04-0270-01 , 3M Catalog# 04-0280-00 , 3M Catalog# 18-0099-68
Specifications
9000 Series, FlexView, ProTop, AP, APC, XL Helmet | |
Velcro™ fasteners | |
Throat Protector |
Speedglas Helmet Accessories | |
FR Cotton |