3M™ Speedglas™ 9000 Series Welding Helmet Accessory, ProTop System

Designed for welders working in areas with overhead hazards

Manufacturer: 3M™ 04001500

 View more versions of this product

Catalog No. 19-160-0304

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Speedglas 9000 Series Welding Helmet
    Product Type Protop System without Welding Filter
    Includes Ratcheted headband
    Weight (Metric) 1.6kg
    Weight (English) 3.53 lb.
    Length (English) Viewing Area 3.58 in.
    Length (Metric) Viewing Area 9.09cm
    For Use With (Application) Ideal for welders working in areas with overhead hazards

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit