3M™ Speedglas™ 9000 Series Welding Helmet Hard Hat ProTop System Accessory, Welding Helm
Manufacturer: 3M™ 04001521SW
 View more versions of this product
Catalog No. 19-160-0319
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) 9000 Series Welding Helmet Hard Hat ProTop System
    Product Type Welding Helmet With Side Windows and 9002X Filter
    For Use With (Application) Ideal for welders working in areas with overhead hazards
    Includes Ratcheted headband, 9002x speedglas™ auto-darkening filter, side windows, two shade 5 side filter, hard hat
    Length (English) Viewing Area 4.09 in.
    Length (Metric) Viewing Area 10.38cm
  • Description & Specifications
Related Products
Product Content Feedback