Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Steri-Strip™ Reinforced Adhesive Skin Closure and Wound Care System

Beathable, microporous material

$85.00

Specifications

Sterility Sterile
For Use With (Application) Enhances patient comfort and less chance of infection, fast application
Color White
Adhesive Hypoallergenic
Latex Free No
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (L x W) Material Quantity Includes Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Material Quantity Includes Price Quantity & Availability  
19-027764
View Documents
3M™
7000002725
3 x 0.25 in. (75 x 6 mm) Rayon 50/Pk. Steri-Strip™ adhesive skin closure
Pack of 50 for $85.00
Case of 4 PK for $307.00
Add to Cart
 
19-086884
View Documents
3M™
7100096926
Dressing: 2.4 x 1.9 in. (60 x 47 mm); Strip: 1.9 x 0.5 in. (47 x 12 mm) Porous Cloth 25/Pk. One reinforced Steri-Strip™ adhesive skin closure and one Tegaderm™ transparent dressing Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
19-027763
View Documents
3M™
7000030208
3 x 0.125 in. (75 x 3 mm) Rayon 50/Pk. Steri-Strip™ adhesive skin closure Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description

Description

  • Proprietary adhesive that does not contain rubber or latex
  • Nonwoven, rayon-backed material reinforced with filaments for strength
  • Adhesive is hypoallergenic
  • Wound Closure System
    • Conbines the cosmetic benefits of the Steri-Strip Skin Closures and the protection of 3M Tegaderm™ Tansparent Dressing into one product
    • 3M Catalog# R1540 , 3M Catalog# R1541 , 3M Catalog# W8512

Specifications

Specifications

Sterile
White
No
Enhances patient comfort and less chance of infection, fast application
Hypoallergenic
Non-invasive
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit