3M™ Supplied-Air Respirator Compatible Breathing Tubes (Loose Fitting Headgear)
Manufacturer: 3M™ 5200160R01
 View more versions of this product
Catalog No. 19-039-702
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) 3M™ air hood system, helmet, headgear and facepiece
    Product Type Breathing Tube
    For Use With (Application) Assembly, machine operation
    Length (Metric) 91.44cm
    Length (English) 36 in.
  • Description & Specifications
Related Products
Product Content Feedback