Learn More
3M SX Eyewear
Sleek, sporty look eye protection
Specifications
Certifications/Compliance | Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007. |
---|---|
Lens Type | Anti-fog |
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog Number | Mfr. No. | Description | Lens Tint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Lens Tint | Price | Quantity & Availability | ||||
19-153-638
|
3M
121350000020 |
Black full frame | Clear |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Available with a full frame, or with a clear-out frame that enhances upward vision
- Wide, contoured seamless lens offers a large, distortion-free field of vision and excellent side protection
- Easy Change system makes lens replacement simple
- Temple lengths adjust for a secure, comfortable fit
- Duralite polycarbonate lens with DX antifog hardcoat protects lens from fog, scratching, and most chemicals
- Lens absorbs 99.9% UV
Compliance
Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
Specifications
Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007. | |
Anti-fog |