Learn More
3M™ T-Series Petroleum Sorbent Sweep
Manufacturer:  3M™ T126
Catalog No. 19-810-260
Add to cart
Description
Description
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
OSHA 29 CFR 1910.132
25 gal.
Petroleum Sorbent Sweep
25 gal., 94.6L
White
94.6L
This sweep incorporates a rope which is attached to one side of the sorbent material, this allows the sorbent to be drawn across an on water spill to recover the petroleum product