Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ 3M™ Thinsulate™-Lined Cold Weather Gloves with Goatskin Palm
This glove is designed to provide excellent hand protection against extreme cold temperatures, complete with a waterproof liner and goatskin palm.
Manufacturer: Wells Lamont™ 894L
Description
The 894 lined goatskin palm glove is designed to provide excellent hand protection against extreme cold temperatures. With 3M™ Thinsulate™ insulation, the 894 glove provides additional warmth and comfort.
- Canvasback wrist with adjustable hook closure creates comfortable, snug fit while keeping out snow, dirt, and cold air
- Thinsulate™ liner
- Waterproof lining
Specifications
ANSI 0 | |
Uncoated | |
ANSI A1 | |
Specific | |
Cold Resistant Gloves | |
Cold Temperature | |
894 |
Cut & Sewn | |
Yellow, Black | |
No Texture | |
Canvas Fabric | |
Large | |
3M™ Thinsulate™ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title