Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M Tie-On Surgical Masks
Flat surgical mask, preferred for its comfort and breathability.
Manufacturer: 3M 7100134016
Description
A tie-on surgical mask with horizontal ties, this medical face mask is ideal for surgical and general-purpose use. Available with or without an attached face shield.
- Flat, with bi-directional pleats and horizontal ties for largely increased facial comfort and fit
- The soft breathable inner lining
- 50 Surgical Masks per box
- 12 Boxes per case
Specifications
EN 14683:2005, ASTM | |
4.67 x 30.22 x 36.57 cm (18.4 x 11.9 x 14.4 in.) | |
14.4 in. | |
Horizontal ties, bi-directional pleats | |
18.4 in. | |
1818 | |
11.9 in. |
Blue | |
>95% (PFE), >99% (BFE) | |
36.57 cm | |
Yes | |
4.67 cm | |
0.1μ | |
30.22 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title