Learn More
3M™ Tour-Guard™ III Visitor's Eyewear
Click to view available options
Product Type:
Small Frame; Dispenser pack
Standard Frame; Dispenser pack
Standard Frame; Polybag pack
Description
- High-impact polycarbonate construction
- Integral sideshields provide wraparound protection
- Large lens fits comfortably—even over prescription eyewear
Order Info
Available in bulk quantities or in handy, ready-to-use wall dispensers.
Compliance
Meets ANSI™ Z87.1-2010 standards.
Specifications
Lens Coating | Uncoated |
Certifications/Compliance | Meets ANSI Z87.1-2003 standards. |
Lens Tint | Clear |
Product Type | Standard Frame; Polybag pack |
No. per Pack | 1 Ea. |
Temple Style | Spatula |
Lens Material | Polycarbonate |