Learn More
3M Ultimate FX FF-400 Series Full Facepiece Respirator
Rugged facepiece designed to provide flexibility in extreme environments
$236.29
Specifications
Connection Type | Bayonet |
---|---|
Product Type | Ultimate Fx Full Facepiece Respirator |
Material | Silicone |
Catalog Number | Mfr. No. | Size | For Use With (Application) | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | For Use With (Application) | Model | Price | Quantity & Availability | |||
19-177-855
|
3M
7000029940 |
Small | Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding | FF-401 |
Each for $236.29
Case of 4 EA for $945.17 |
|
19-177-857
|
3M
7100001847 |
Large | Assembly, cleaning, grinding, machine operation, painting, remediation, welding | FF-403 |
Each for $236.29
Case of 4 EA for $945.17 |
|
19-177-856
|
3M
7000002284 |
Medium | Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding | FF-402 |
Each for $236.29
Case of 4 EA for $945.17 |
|
Description
- Large lens with superior peripheral vision delivers maximum comfort with a wide field of view
- Exclusive Scotchgard coating on lens causes some liquids to bead up on the surface so they can be wiped off easily
- Durable, permanently attached gaskets for easy maintenance and longer life
- Low-profile bayonet design improves balance and offers better weight distribution
- Ideal for tight work environments
- Durable head harness with six-strap configuration fits securely
- Comfort cradle positions respirator comfortably on head
- Soft, silicone nose cup and faceseal fits broad range of face shapes and sizes
- Cool Flow exhalation valve allows easier breathing, reduces heat and moisture buildup
- 3M Catalog# FF-401 , 3M Catalog# FF-402 , 3M Catalog# FF-403
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Specifications
Bayonet | |
Ultimate Fx Full Facepiece Respirator | |
Silicone |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.