Learn More
3M™ Versaflo M-400 Series PAPR Outer Shroud
Outer shrouds are replacement parts for use with M-400 helmets.
Supplier: 3M™ 7000127685
This item is not returnable.
View return policy
Description
- The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
- 3M Catalog# M-445, M-446, M-447, M-448
Specifications
Outer Shroud, Comfort | |
Cleaning, electrical, facility maintenance, painting, sanding, welding | |
M-400 helmet | |
M-446 |