3M™ Versaflo™ PAPR System Filters and Cartridges

Compatible with powered air purifying respirators in the TR-300 and TR-600 Series

Manufacturer: 3M™ TR6590N

 View more versions of this product

Catalog No. 19-268-015

  Check Availability
Add to Cart

    Specifications

    Quantity 10 Pack
    For Use With (Equipment) TR-600 PAPR,TR-600 PAPR
    Product Type Cartridge
    For Use With (Application) Pharmaceutical
    Certifications NIOSH
    Filter Type Combination HE Particulate/Multi-gas (MG)
    Model TR-6590N
    Color Magenta/Olive

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit