3M™ Versaflo™ PAPR System Filters and Cartridges

Compatible with powered air purifying respirators in the TR-300 and TR-600 Series

Manufacturer: 3M™ TR-3710N-40

 View more versions of this product

Catalog No. 19-039-573

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Quantity 10 Pack
    For Use With (Equipment) TR-300 PAPR,TR-300 PAPR
    Product Type High Efficiency Filter
    Certifications NIOSH
    Filter Type Particulate
    Model TR-3710N-40
    Color Magenta

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit