Learn More
3M™ Versaflo TR-300 Series PAPR Headcovers
Easiest way to select a complete system for hospital, healthcare, and pharmaceutical applications where an S-133, APF 25 headcover is preferred
Manufacturer:  3M™ TR300HKS
 View more versions of this product
Catalog No. 19-039-554
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Before use, the wearer must read and understand the USER Instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting all the requirements of OSHA 1910.134 including training, fit testing and medical evaluation. In Canada, CSA standards Z94.4 requirements must be met and/or requirements of the applicable jurisdiction, as appropriate. Misuse may result in sickness or death.
Description
Description
Kit Components
  • S-133 Headcover
  • TR-307N PAPR with Easy Clean Belt
  • BT-30 Breathing Tube
  • Charger
  • TR-3710N filter
  • Storage bag
  • Sizes: Small/Medium and Medium/Large
Specifications
Specifications
Gray
Small/Medium
Tr-300 series powered air purifying respirator
Back
4.3 kg
PAPR Kit
Pharmaceutical, healthcare and food and beverage industries
Battery, vinyl belt, charger, filter, breathing tube with cover, headcover and carrying/storage bag
9.5 lb.