Learn More
3M™ Versaflo TR-300 Series PAPR Headcovers
Easiest way to select a complete system for hospital, healthcare, and pharmaceutical applications where an S-133, APF 25 headcover is preferred
Manufacturer:  3M™ TR300HKL
 View more versions of this product
Catalog No. 19-039-553
  Check Availability
Add to cart
Alert:
Before use, the wearer must read and understand the USER Instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting all the requirements of OSHA 1910.134 including training, fit testing and medical evaluation. In Canada, CSA standards Z94.4 requirements must be met and/or requirements of the applicable jurisdiction, as appropriate. Misuse may result in sickness or death.
Specifications Description & Specifications
Specifications
Gray
PAPR Kit
Battery, vinyl belt, charger, filter, breathing tube with cover, headcover and carrying/storage bag
Medium/Large
9.5 lb.
Tr-300 series powered air purifying respirator
Pharmaceutical, healthcare and food and beverage industries
Back
4.3kg