Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Versaflo™ TR-600 Series PAPR Belts and Extenders
For use with Versaflo™ TR-600 Series PAPR
Supplier: 3M™ 7100035833
This item is not returnable.
View return policy
Description
TR-626 High Durability Belt
- Comfortable belt due to ergonomic design
- Durable construction and easy to use
- Leather leads make it suitable for grinding
- Part of TR-613N and TR-616N assemblies, TR-600-HIK Ready-to-Use Kit
- Accommodates waist sizes up to 52 in.
- Applications: Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
- Color: Gray TR-626X Belt Extender
- Comfortable due to ergonomic design
- Easy to use extender
- Enables TR-626 belt to be worn comfortably, when additional belt length is needed
- Multiple TR-626X Belt Extenders may be connected in series
- Applications: Healthcare, Shutdown, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
- Color: Gray TR-627 Easy Clean Belt
- Comfortable due to ergonomic design
- Easy to use belt
- Smooth surfaces make it easier to clean
- Part of TR-614N and TR-617N assemblies, TR-600-ECK Ready-to-Use Kit
- Accommodates waist sizes up to 52 in.
- Applications: Healthcare, Pharmaceutical, Pesticide Application, Animal processing, Laboratories, Nuclear energy, Cosmetics, Pharmaceutical research and production, Recycling, Dry Foods
- Color: Gray TR-627X Belt Extender
- Comfortable due to ergonomic design
- Easy to use extender
- Enables TR-627 belt to be worn comfortably, when additional belt length is needed
- Multiple TR-627X Belt Extenders may be connected in series
- Applications: Healthcare, Pharmaceutical, Pesticide Application, Animal processing, Laboratories, Nuclear energy, Cosmetics, Pharmaceutical research and production, Recycling, Dry Foods
- Color: Gray
- 3M Catalog# TR-626, TR-626X, TR-627, TR-627X
Certifications
NIOSH Approved
Specifications
Gray | |
Belt | |
Versaflo™ TR-600 Series PAPR | |
1 Ea. |
Reusable | |
Healthcare, Pharmaceutical, Pesticide Application, Animal Processing, Laboratories, Nuclear Energy, Cosmetics, Pharmaceutical Research and Production, Recycling, Dry Foods | |
TR-627 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction