Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ TR-600 Series Filter Cover

For use with Versaflo™ TR-600 Series PAPR systems.

Specifications

Certifications/Compliance NIOSH
Color Gray
Product Type Filter Cover
Quantity 1 Ea.
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
19-268-033
Required Accessories View Documents
3M™
7100021251
TR-6700 and TR-600 Respirators, TR-6600 Prefilter, and TR-662 Spark Arrestor
Case for $25.26
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-268-026
Required Accessories View Documents
3M™
7100021252
TR-6500 and TR-600 Respirators, TR-6600 Prefilter, and TR-662 Spark Arrestor
Case for $31.83
N/A
Edge
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Filter cover helps protect cartridge from physical damage.
  • Easy to use cover enables use of prefilters and/or spark arrestors.
  • Allows PAPR to be worn through decontamination showers or rain.
  • Also used to hold TR-6600 Prefilter or TR-662 Spark Arrestor in place.
  • Recommended Industries include healthcare, pharmaceutical, heavy industrial, manufacturing.
Specifications

Specifications

NIOSH
Gray
Filter Cover
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.