3M™ Versaflo™ Units and Assemblies for TR-300 Series PAPRs

Provide protection against particulates and certain nuisance odors

Manufacturer: 3M™ TR306N

 View more versions of this product

Catalog No. 19-039-558

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Tr-300 series powered air purifying respirator,Tr-300 series powered air purifying respirator
    Product Type Powered Air Purifying Respirator Assembly
    For Use With (Application) Assembly and mechanical, cleaning, cutting, drilling, general maintenance, grinding, metal repair, painting, powder coating, sawing
    Includes Easy clean belt, high capacity battery, he filter and single-station charger
    Weight (Metric) 1.133kg
    Weight (English) 2.5 lb.
    Filter Type HE Filter
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit