Learn More
3M™ VFlex™ Health Care Particulate Respirator and Surgical Mask
Manufacturer:  3M™ 7000148531
 View more versions of this product
Catalog No. 19-177-911
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This disposable N95 Health Care Particulate Respirator and Surgical Mask is designed to help provide quality, reliable respiratory protection for health care workers. CDC guidelines state N95 respirators may be used for M. tuberculosis exposure control.* As a disposable particulate respirator, it is intended to help reduce wearer exposure to certain airborne particles including those generated by electrocautery, laser surgery, and other powered medical instruments. As a surgical mask, it is designed to be fluid resistant to splash and spatter of blood and other infectious materials. 3M uses a variety of innovative technologies and features to help you meet your respiratory protection and comfort needs. This economical flatfold respirator has the following features:
  • NIOSH Approved N95
  • FDA 510K cleared
  • Fluid Resistant as tested per ASTM F1862
  • Meets CDC guidelines for M. tuberculosis exposure control
  • 3M™ Advanced Electrostatic Media (AEM) - Highly charged microfibers enhance the capture of airborne particles while allowing you to breathe easier
  • Pleats help increase surface area for ease of breathing
  • Pleats flex with mouth movement while talking
  • Adjustable noseclip helps provide a custom and secure seal
  • Available in 2 sizes to fit a broad range of faces
  • Tabs uniquely designed for positioning the respirator on the face
Specifications
Specifications
Yes
White
1805S
ASTM F1862, NIOSH N95, FDA 510K
Small