Learn More
3M™ VFlex™ N95 Health Care Particulate Respirator and Surgical Mask
This particulate respirator and surgical mask helps provide respiratory protection against certain airborne particles. The folded respirator expands for a spacious feel and the pleats help increase surface area for ease of breathing.
Specifications
Certifications/Compliance ASTM F1862, NIOSH N95, FDA 510K
Color White
Disposable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-177-910
View Documents
3M™
1805
Standard This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-177-911
View Documents
3M™
7000148531
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
This disposable N95 Health Care Particulate Respirator and Surgical Mask is designed to help provide quality, reliable respiratory protection for health care workers. CDC guidelines state N95 respirators may be used for M. tuberculosis exposure control.* As a disposable particulate respirator, it is intended to help reduce wearer exposure to certain airborne particles including those generated by electrocautery, laser surgery, and other powered medical instruments. As a surgical mask, it is designed to be fluid resistant to splash and spatter of blood and other infectious materials. 3M uses a variety of innovative technologies and features to help you meet your respiratory protection and comfort needs. This economical flatfold respirator has the following features:
  • NIOSH Approved N95
  • FDA 510K cleared
  • Fluid Resistant as tested per ASTM F1862
  • Meets CDC guidelines for M. tuberculosis exposure control
  • 3M™ Advanced Electrostatic Media (AEM) - Highly charged microfibers enhance the capture of airborne particles while allowing you to breathe easier
  • Pleats help increase surface area for ease of breathing
  • Pleats flex with mouth movement while talking
  • Adjustable noseclip helps provide a custom and secure seal
  • Available in 2 sizes to fit a broad range of faces
  • Tabs uniquely designed for positioning the respirator on the face
Specifications
Specifications
ASTM F1862, NIOSH N95, FDA 510K
White
Yes