Learn More
3M™ Virtua Max Eyewear
Click to view available options
Lens Tint:
Clear
Gray
Lens Coating:
Anti-fog
Description
- Clear anti-fog lens or Indoor/outdoor gray anti-fog lens
- Clear frame
- Large lens size
- Meets the requirements of CSA Z94.3-2007 and the High Impact Requirements of ANSI Z87.1-2003
- Lightweight, unisex protective eyewear
- High wraparound coverage
- Wraparound design provides extended eye protection and unobstructed viewing
- Contoured temples fit comfortably and snugly
- Polycarbonate lenses absorb 99.9% UVA and UVB
- 3M Catalog# 11511-00000-20
Specifications
Lens Tint | Clear |
Lens Material | Polycarbonate |