3M™ Virtua™ Slim Eyewear
Designed specifically for smaller facial profiles
Manufacturer: 3M™ 117560000020
 View more versions of this product
Catalog No. 19-810-018
  Check Availability
Add to Cart
Alert
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
    Specifications
    Lens Coating Hardcoated
    Temple Color Gray
    Certifications/Compliance Meets ANSI Z87.1-2010 standard.
    Lens Material Polycarbonate
    Lens Tint Gray
  • Description & Specifications
Related Products
Product Content Feedback