Learn More
3M™ Welding Helmet 10 Parts and Accessories
For use with 3M Welding Helmet 10
Supplier:  3M™ 165005
 View more versions of this product
Catalog No. 19-031-141
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Replacement Parts for 3M™ Welding Helmet 10
  • Sweatband
  • Mounting Hardware without Headband
  • Headband and Mounting Hardware
3M™ Outside Protection Plate 10
  • Designed to protect the filter from spark and spatter
  • Failure to use an outside protection plate with the filter voids the warranty if spark or spatter contaminates the glass
  • Size: 90 x 110 mm
3M™ Auto-Darkening Filter 10V
  • Good for Stick and MIG welding processes
  • Offers user selectable dark shades 10 through 12 and two sensitivity adjustments
Specifications
Specifications
3M Welding Helmet 10
Replacement Headband with mounting hardware
Product Suggestions
Product Suggestions