Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R)-(+)-3-Aminopyrrolidine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
116183-82-5
C4H10N2
86.138
MFCD00143190
NGXSWUFDCSEIOO-SCSAIBSYSA-N
1519352
(3R)-pyrrolidin-3-amine
C1CNCC1N
116183-82-5
C4H10N2
86.138
MFCD00143190
NGXSWUFDCSEIOO-SCSAIBSYSA-N
1519352
(3R)-pyrrolidin-3-amine
C1CNCC1N
Quantity 25g
Color Yellow
UN Number 2735
Formula Weight 86.14
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (3R)-(+)-3-Aminopyrrolidine