Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
[(3R)-3-Hydroxyoctadecanoyl]-L-carnitine, ≥94.0% (HPLC), MilliporeSigma™ Supelco™
$418.00
Specifications
CAS | 1469901-05-0 |
---|---|
Type | Analytical Standard |
Linear Formula | C25H49 NO5 |
Synonym | (2 R)-3-Carboxy-2-[[(3 R)-3-hydroxy-1-oxooctadecyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt; [(R)-3-Hydroxystearoyl]-L-carnitine |
Formula Weight | 443.66 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-3154
|
MilliporeSigma Supelco
5241310MG |
10 mg |
Each of 1 for $418.00
|
|
|||||
Specifications
1469901-05-0 | |
C25H49 NO5 | |
443.66 | |
Analytical Standard |
Analytical Standard | |
(2 R)-3-Carboxy-2-[[(3 R)-3-hydroxy-1-oxooctadecyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt; [(R)-3-Hydroxystearoyl]-L-carnitine | |
≥94.0% (HPLC) |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title