Learn More
[(3R)-3-Hydroxytetradecanoyl]-L-carnitine, ≥90% (HPLC), MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 5200250MG
 View more versions of this product
Catalog No. 11-101-3151
Add to cart
Description
Description
Specifications
Specifications
1469901-02-7
(2 R)-3-Carboxy-2-[[(3 R)-3-hydroxy-1-oxotetradecyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt
≥90% (HPLC)
50 mg
C21H41 NO5
387.55
Analytical Standard
Analytical Standard
Safety and Handling
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Recommended Storage : -20°C