Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
163439-82-5
C11H15NO2
193.24
MFCD01073894
QJRIUWQPJVPYSO-GHMZBOCLSA-N
3r,4r-1-benzylpyrrolidine-3,4-diol,3r,4r---1-benzyl-3,4-pyrrolidindiol,3r,4r---1-benzyl-3,4-pyrrolidinediol,3,4-pyrrolidinediol, 1-phenylmethyl-, 3r,4r,pubchem14982,trans-3,4-dihydroxy-n-benzylpyrrolidine,3r,4r-1-benzyl-3,4-pyrrolidinediol,3r-1-benzylpyrrolidine-3beta,4alpha-diol,3r,4r-rel-1-phenylmethyl-3,4-pyrrolidinediol,3,4-pyrrolidinediol,1-phenylmethyl-, 3r,4r
7021491
(3R,4R)-1-benzylpyrrolidine-3,4-diol
C1C(C(CN1CC2=CC=CC=C2)O)O
163439-82-5
C11H15NO2
193.24
MFCD01073894
QJRIUWQPJVPYSO-GHMZBOCLSA-N
3r,4r-1-benzylpyrrolidine-3,4-diol,3r,4r---1-benzyl-3,4-pyrrolidindiol,3r,4r---1-benzyl-3,4-pyrrolidinediol,3,4-pyrrolidinediol, 1-phenylmethyl-, 3r,4r,pubchem14982,trans-3,4-dihydroxy-n-benzylpyrrolidine,3r,4r-1-benzyl-3,4-pyrrolidinediol,3r-1-benzylpyrrolidine-3beta,4alpha-diol,3r,4r-rel-1-phenylmethyl-3,4-pyrrolidinediol,3,4-pyrrolidinediol,1-phenylmethyl-, 3r,4r
7021491
(3R,4R)-1-benzylpyrrolidine-3,4-diol
C1C(C(CN1CC2=CC=CC=C2)O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Brown to White
Melting Point 95.0°C to 101.0°C
Specific Rotation − 32.00
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 193.24
Physical Form Powder, Crystals, and/or Chunks
Percent Purity 97%
Chemical Name or Material (3R, 4R)-(-)-1-Benzyl-3, 4-pyrrolidindiol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning