Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,6R)-3,6-Octanediol, 95%, (98% ee), Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
129619-37-0
C8H18O2
146.23
MFCD01631087
BCKOQWWRTRBSGR-HTQZYQBOSA-N
3r,6r-3,6-octanediol,3r,6r-octane-3,6-diol,3,6-octanediol, 3r,6r,3r,3r-3,6-octadiol,3r,6r-3,6-dihydroxyoctane,3r,6r---3,6-octanediol
2734541
(3R,6R)-octane-3,6-diol
CCC(CCC(CC)O)O
129619-37-0
C8H18O2
146.23
MFCD01631087
BCKOQWWRTRBSGR-HTQZYQBOSA-N
3r,6r-3,6-octanediol,3r,6r-octane-3,6-diol,3,6-octanediol, 3r,6r,3r,3r-3,6-octadiol,3r,6r-3,6-dihydroxyoctane,3r,6r---3,6-octanediol
2734541
(3R,6R)-octane-3,6-diol
CCC(CCC(CC)O)O
Packaging Glass Bottle
Infrared Spectrum Authentic
Color White
Enantiomeric Excess 97.5% min. (GC)
Quantity 1g
Formula Weight 146.23
Physical Form Needle-Like Crystals
Percent Purity ≥94%
Chemical Name or Material (3R, 6R)-3, 6-Octanediol, 95%, (98% e.e.)