Alfa Aesar™ (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one, 98%

Manufacturer: Alfa Aesar™ H2716406

Quantity 5g

 View more versions of this product

Catalog No. AAH2716406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97538-67-5
    Molecular Formula C7H8Cl3NO2
    Formula Weight 244.51
    MDL Number MFCD08274464
    Synonym (2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one; L-Proline-chloral condensation product
    Chemical Name or Material (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one

    Specifications

    Formula Weight 244.51
    Molecular Formula C7H8Cl3NO2
    MDL Number MFCD08274464
    CAS 97538-67-5
    Synonym (2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one; L-Proline-chloral condensation product
    Melting Point 109°C
    Quantity 5g
    Optical Rotation −2.9° (c= 1 in Chloroform)
    Chemical Name or Material (3R-cis)-Tetrahydro-3-trichloromethyl-1H,3H-pyrrolo[1,2-c]oxazol-1-one
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit