Alfa Aesar™ (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol, 98%

Manufacturer: Alfa Aesar™ H2813706

Quantity 5g

 View more versions of this product

Catalog No. AAH2813706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 105812-81-5
    Molecular Formula C13H18FNO
    Formula Weight 223.29
    MDL Number MFCD06658161
    UN Number UN3077
    Synonym (3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
    Chemical Name or Material (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol

    Specifications

    Formula Weight 223.29
    Molecular Formula C13H18FNO
    MDL Number MFCD06658161
    CAS 105812-81-5
    UN Number UN3077
    Synonym (3S,4R)-4-(4-Fluorophenyl)-3-hydroxymethyl-1-methylpiperidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P261-P280b-P262-P273-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves and eye protection.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 93° to 98°C
    Quantity 5g
    Optical Rotation −34° (c=1 in Methanol)
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol
    Assay 98%

    GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P261-P280b-P262-P273-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves and eye protection.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit